Learn More
Cayman Chemical Ravuconazole 10mg
SDP
Catalog No. 502967924
Encompass_Preferred
An orally available azole fungicide that potently inhibits the growth of a wide range of fungi (MICs range from 25 to 780 ng/ml); inhibits CYP isoforms that are involved in ergosterol biosynthesis, interfering with the generation of the fungal and protozoan cell membranes
Catalog No. 50-296-7924 Supplier Cayman Chemical Supplier No. 1875010 MG
An orally available azole fungicide that potently inhibits the growth of a wide range of fungi (MICs range from 25 to 780 ng/ml); inhibits CYP isoforms that are involved in ergosterol biosynthesis, interfering with the generation of the fungal and protozoan cell membranes