Learn More
ATCC Mus musculus
Supplier:  ATCC TIB71/DEL
Encompass
RAW 264.7 is a macrophage cell line that was established from a tumor in a male mouse induced with the Abelson murine leukemia virus.
Catalog No. 50-188-870FP