Learn More
ATCC RAW 264.7 gamma NO (-); Macrophage; Mouse (Mus musculus)
Catalog No. 502383223
Encompass
RAW 264.7 gamma NO(-) was derived from the RAW 264.7 (see ATCC TIB-71) mouse monocyte/macrophage cell line Unlike the parental line, RAW 264.7 gamma NO(-) does not produce nitric oxide upon treatment with interferon gamma alone, but requires LPS for full activation (the iNOS promoter linked to a luciferase reporter gene is also unresponsive to IFN- alone). This cell line was deposited by SW Russell and can be used in assay development.
Catalog No. 50-238-3223 Supplier ATCC Supplier No. CRL2278
RAW 264.7 gamma NO(-) was derived from the RAW 264.7 (see ATCC TIB-71) mouse monocyte/macrophage cell line Unlike the parental line, RAW 264.7 gamma NO(-) does not produce nitric oxide upon treatment with interferon gamma alone, but requires LPS for full activation (the iNOS promoter linked to a luciferase reporter gene is also unresponsive to IFN- alone). This cell line was deposited by SW Russell and can be used in assay development.