Learn More
ATCC RAW 264.7; Macrophage; Mouse (Mus musculus)
Supplier:  ATCC TIB71
Encompass
RAW 264.7 is a macrophage cell line that was established from a tumor in a male mouse induced with the Abelson murine leukemia virus.
Catalog No. 50-238-2304