Learn More
Invivogen RAW-Lucia™ ISG-KO-MAVS Cells, MAVS Knockout IRF-Lucia Luciferase Reporter Cells, 3-7 x 10e6 cells
SDP
Catalog No. NC3450191
Encompass
RAW-Lucia™ ISG-KO-MAVS cells were generated from RAW-Lucia™ ISG cells through the stable knockout of the MAVS gene. These cells derive from the murine RAW 264.7 macrophage cell line, which has been reported to express many pattern recognition receptors (PRRs), including the dsRNA sensors MDA-5 [1] and RIG-I [2, 3] along with their adaptor protein MAVS [3].RAW-Lucia™ ISG-KO-MAVS and RAW-Lucia™ ISG cells can be used to study the role of MAVS by monitoring of IRF-induced Lucia luciferase activity. They express the gene for secreted Lucia luciferase under the control of an IFN-inducible IFN-stimulated genes 54 (ISG54) promoter enhanced by a multimeric IFN-stimulated response elements (ISRE). The levels of IRF-induced Lucia in the cell culture supernatant can be easily monitored using QUANTI-Luc™ 4 Lucia/Gaussia, a Lucia and Gaussia luciferase detection reagent.RAW-Lucia™ ISG-KO-MAVS cells are resistant to Zeocin®.
Catalog No. NC3450191 Supplier Invivogen Supplier No. RAWLKOMAVS
Add to Cart
Add to Cart
RAW-Lucia™ ISG-KO-MAVS cells were generated from RAW-Lucia™ ISG cells through the stable knockout of the MAVS gene. These cells derive from the murine RAW 264.7 macrophage cell line, which has been reported to express many pattern recognition receptors (PRRs), including the dsRNA sensors MDA-5 [1] and RIG-I [2, 3] along with their adaptor protein MAVS [3].RAW-Lucia™ ISG-KO-MAVS and RAW-Lucia™ ISG cells can be used to study the role of MAVS by monitoring of IRF-induced Lucia luciferase activity. They express the gene for secreted Lucia luciferase under the control of an IFN-inducible IFN-stimulated genes 54 (ISG54) promoter enhanced by a multimeric IFN-stimulated response elements (ISRE). The levels of IRF-induced Lucia in the cell culture supernatant can be easily monitored using QUANTI-Luc™ 4 Lucia/Gaussia, a Lucia and Gaussia luciferase detection reagent.RAW-Lucia™ ISG-KO-MAVS cells are resistant to Zeocin®.