Learn More
Aviva Systems Biology Corp RAX Antibody - N-terminal region (ARP32164_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32164_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAX. Alternate names: RX, MCOP3ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Rat
Catalog No. 50-014-54470