Learn More
Medchemexpress LLC HY-12796A 5mg Medchemexpress, Raxatrigine hydrochloride CAS:934240-31-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12796A5MG
Encompass_Preferred
Medchemexpress, HY-12796A 5mg Raxatrigine hydrochloride CAS:934240-31-0 Raxatrigine hydrochloride (GSK-1014802 hydrochloride) is a novel small molecule state-dependent sodium channel blocker; Nav1.7 sodium channel inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1146