Learn More
Medchemexpress LLC Raxatrigine hydrochloride | 934240-30-9 | 99.85% | 10 MG
SDP
Catalog No. 5000339271
Encompass_Preferred
Raxatrigine hydrochloride is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. It is intended for research use only.
  • High purity
  • Detailed solubility information for in vitro and in vivo applications
  • Orphan-drug designation
  • Access to data sheet, certificate of analysis, safety data sheet, and handling instructions
  • Contextual clinical trial information available
  • Useful for neurological disease research
  • Targets sodium channel
Catalog No. 50-003-39271 Supplier Medchemexpress LLC Supplier No. HY12796A10MG
Raxatrigine hydrochloride is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. It is intended for research use only.
  • High purity
  • Detailed solubility information for in vitro and in vivo applications
  • Orphan-drug designation
  • Access to data sheet, certificate of analysis, safety data sheet, and handling instructions
  • Contextual clinical trial information available
  • Useful for neurological disease research
  • Targets sodium channel