Learn More
Medchemexpress LLC Raxatrigine hydrochloride | 934240-31-0 | 99.1% | 1 ML
SDP
Catalog No. 5000340761
Encompass_Preferred
Raxatrigine hydrochloride, also known as Vixotrigine hydrochloride, is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. This compound is intended for research use only.
  • Inhibits Nav1.7 sodium channel
  • Acts as a state-dependent sodium channel blocker
  • Soluble in DMSO and water
  • Compatible with various in vivo protocols
  • Specific storage recommendations for solid and solution forms
Catalog No. 50-003-40761 Supplier Medchemexpress LLC Supplier No. HY12796A10MM/1ML
Raxatrigine hydrochloride, also known as Vixotrigine hydrochloride, is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. This compound is intended for research use only.
  • Inhibits Nav1.7 sodium channel
  • Acts as a state-dependent sodium channel blocker
  • Soluble in DMSO and water
  • Compatible with various in vivo protocols
  • Specific storage recommendations for solid and solution forms