Learn More
Medchemexpress LLC Raxatrigine (hydrochloride) | 934240-31-0 | 99.1% | 100 MG
SDP
Catalog No. 5000340760
Encompass_Preferred
Raxatrigine hydrochloride, also known as Vixotrigine hydrochloride or GSK-1014802 hydrochloride, is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. It is intended for research use only and received orphan-drug designation from the US Food and Drug Administration in July 2013.
  • Novel small molecule state-dependent sodium channel blocker
  • Nav1.7 sodium channel inhibitor
  • Received orphan-drug designation from US FDA
  • Potential in the treatment of cognitive symptoms of schizophrenia
  • Not sold to patients; for research use only
  • Purity of 99.1%
Catalog No. 50-003-40760 Supplier Medchemexpress LLC Supplier No. HY12796A100MG
Raxatrigine hydrochloride, also known as Vixotrigine hydrochloride or GSK-1014802 hydrochloride, is a novel small molecule state-dependent sodium channel blocker and a Nav1.7 sodium channel inhibitor. It is intended for research use only and received orphan-drug designation from the US Food and Drug Administration in July 2013.
  • Novel small molecule state-dependent sodium channel blocker
  • Nav1.7 sodium channel inhibitor
  • Received orphan-drug designation from US FDA
  • Potential in the treatment of cognitive symptoms of schizophrenia
  • Not sold to patients; for research use only
  • Purity of 99.1%