Learn More
Aviva Systems Biology Corp RAXL1 Antibody - N-terminal region (ARP36080_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36080_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RAXL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAXL1. Alternate names: QRX, ARMD6, RAXL1, CORD11ApplicationWB. Species reactivity: Cow, Dog, Human, Pig
Catalog No. 50-014-56006