Learn More
Raybiotech Inc RayBright® LIVE V500 100 TESTS
Catalog No. NC2902807
Encompass_Preferred
RayBright® Live V500 is an amine-reactive fluorescent dye that binds both live and dead cells, with a much brighter signal in dead cells than living cells which refuse the dye while dead cells are stained both on the cell surface and intracellularly. This dye can be used to discriminate live and dead cells in flow cytometry analysis and is supplied frozen in DMSO solution at a pre-titrated concentration. End users are recommended to further determine the optimal concentration for their specific cell types
Catalog No. NC2902807 Supplier Raybiotech Inc Supplier No. 13709003100
RayBright® Live V500 is an amine-reactive fluorescent dye that binds both live and dead cells, with a much brighter signal in dead cells than living cells which refuse the dye while dead cells are stained both on the cell surface and intracellularly. This dye can be used to discriminate live and dead cells in flow cytometry analysis and is supplied frozen in DMSO solution at a pre-titrated concentration. End users are recommended to further determine the optimal concentration for their specific cell types