Learn More
Raylabcon Vinyl Microscope Slides
SDPSCI_ED
Catalog No. S17526
Change view
Click to view available options
Coating:
Scratch-resistant
Catalog No. Coating
S17526 Scratch-resistant
1 options
Catalog No. S17526 Supplier Raylabcon Supplier No. 20 30 5201
Add to Cart
Add to Cart
Raylabcon Vinyl Microscope Slides
Specifications
Length (Metric) 75 mm
Thickness (Metric) 0.0180 to 0.022 mm
Width (Metric) 25 mm
Material Vinyl
Color Clear
Coating Scratch-resistant
Quantity 144/Pk.