Learn More
MSC AbilityOne Rayon Roll Wiping Towel, 36 in x 250 yds, Absorbent, Low-Lint
Catalog No. 502839199
Encompass
This rayon roll wiping towel is a low-lint, highly absorbent, soft, all-purpose, and marine-degradable wipe. It is designed to quickly and effectively soak up spills and is perfect for wiping down equipment. It can be easily cut to any desired length.
  • Low-lint and highly absorbent
  • Soft, all-purpose wipe
  • Marine-degradable
  • Can be easily cut to desired length
  • Effectively soaks up spills
  • Perfect for wiping down equipment
  • Recommended for Navy replacement for baled rags and terry cloth
  • Biodegradable
  • Features PRIME (Plastics Removal in the Marine Environment) recommendation
  • White color
Catalog No. 50-283-9199 Supplier MSC Supplier No. 70871272
This rayon roll wiping towel is a low-lint, highly absorbent, soft, all-purpose, and marine-degradable wipe. It is designed to quickly and effectively soak up spills and is perfect for wiping down equipment. It can be easily cut to any desired length.
  • Low-lint and highly absorbent
  • Soft, all-purpose wipe
  • Marine-degradable
  • Can be easily cut to desired length
  • Effectively soaks up spills
  • Perfect for wiping down equipment
  • Recommended for Navy replacement for baled rags and terry cloth
  • Biodegradable
  • Features PRIME (Plastics Removal in the Marine Environment) recommendation
  • White color