Learn More
World Precision Instrument Razor Blade Holder, 12 cm
SDP
Catalog No. 50822253
Encompass
This reusable surgical instrument lets you securely hold and manipulate disposable scalpel blades during surgical procedures. The unique concave/convex jaw prevents the blades from slipping, and the locking mechanism on the long handle secures the blade in place. The scissor-like handles give excellent control and leverage for the user. This scalpel handle may be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. This scalpel handle is available in surgical grade stainless steel or our premium German steel.
Catalog No. 50-822-253 Supplier World Precision Instrument Supplier No. 14134
This reusable surgical instrument lets you securely hold and manipulate disposable scalpel blades during surgical procedures. The unique concave/convex jaw prevents the blades from slipping, and the locking mechanism on the long handle secures the blade in place. The scissor-like handles give excellent control and leverage for the user. This scalpel handle may be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect, and sterilize your surgical instruments promptly after each use to ensure a long life for your surgical tools. This scalpel handle is available in surgical grade stainless steel or our premium German steel.