Learn More
Electron Microscopy Sciences Double Edge Non-Coated Blade, 250/box
SDP
Catalog No. 50366009
Encompass_Preferred
Stainless Steel "PTFE" coated. PERSONNA brand .004" thick, wrapped individually. Washed and degreased reducing foreign material. Clean room classified.
Catalog No. 50-366-009 Supplier Electron Microscopy Sciences Supplier No. 71998
Stainless Steel "PTFE" coated. PERSONNA brand .004" thick, wrapped individually. Washed and degreased reducing foreign material. Clean room classified.