Learn More
Med Vet International McKesson Twin-Blade Disposable Razor, Blue, 16-RZ50, 100/Case
Catalog No. 503043544
Encompass_Preferred
Razor McKesson Twin Blade Disposable The McKesson disposable razor is single-use to protect patients and caregivers from cross-contamination A fixed head on a curved arm provides added precision Twin blades sit in an arch housing to achieve a close, smooth shave A ribbed handle offers a secure, comfortable grip A flip-off blade cover provides safety during handling and storage of the fixed head razor Comes without the moisturing strip
Catalog No. 50-304-3544 Supplier Med Vet International Supplier No. 474704CS
Razor McKesson Twin Blade Disposable The McKesson disposable razor is single-use to protect patients and caregivers from cross-contamination A fixed head on a curved arm provides added precision Twin blades sit in an arch housing to achieve a close, smooth shave A ribbed handle offers a secure, comfortable grip A flip-off blade cover provides safety during handling and storage of the fixed head razor Comes without the moisturing strip