Learn More
Electron Microscopy Sciences GEM Runner™ 2 Single Edge Blade, 2 facet
SDP
Supplier:  Electron Microscopy Sciences 7198518
Encompass_Preferred
GEM 2 facet single-edge blades work with the Gem Runner Pop Up Dispenser. Made of carbon steel.
Material: Carbon Steel
Type: 2 facet
Degreased: Yes
Catalog No. 50-192-8060