Learn More
Electron Microscopy Sciences GEM Runner™ 3 Single Edge Blade, 3 facet
SDP
Catalog No. 501928056
Encompass_Preferred
GEM 3 facet single-edge blades work with the Gem Runner Pop Up Dispenser. Made of carbon steel.
Material: Carbon Steel
Type: 3 facet
Degreased: Yes
Catalog No. 50-192-8056 Supplier Electron Microscopy Sciences Supplier No. 7198513
GEM 3 facet single-edge blades work with the Gem Runner Pop Up Dispenser. Made of carbon steel.
Material: Carbon Steel
Type: 3 facet
Degreased: Yes