Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Rb aHu IgM Anti Secondary

Supplier:  Cytek Biosciences Inc 708075M001

Encompass_Preferred

Product Name: Unconjugated Rabbit Anti-Human IgM
Product Class: Antibody
Type: Secondary
Product Name: Unconjugated Rabbit Anti-Human IgM
Cytek Catalog Number: 70-8075-M001
Marker: IgM
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Conjugation: Unconjugated
Size: 1 mg

Catalog No. 50-000-87040


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.