Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Rb aHu IgM Anti Secondary

Catalog No. 5000087307
Encompass_Preferred

Product Name: Biotin Rabbit Anti-Human IgM
Product Class: Antibody
Type: Secondary
Product Name: Biotin Rabbit Anti-Human IgM
Cytek Catalog Number: 30-8075-M001
Marker: IgM
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Conjugation: Biotin
Size: 1 mg

Catalog No. 50-000-87307 Supplier Cytek Biosciences Inc Supplier No. 308075M001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: Biotin Rabbit Anti-Human IgM
Product Class: Antibody
Type: Secondary
Product Name: Biotin Rabbit Anti-Human IgM
Cytek Catalog Number: 30-8075-M001
Marker: IgM
Target Species: Human
Reported Cross Reactivity: Human
Clonality: Polyclonal
Host Species: Rabbit
Conjugation: Biotin
Size: 1 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.