Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc Rb aMu IgG Anti Secondary

Supplier:  Cytek Biosciences Inc 308077M002

Encompass_Preferred

Product Name: Biotin Rabbit Anti-Mouse IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: Biotin Rabbit Anti-Mouse IgG (H&L)
Cytek Catalog Number: 30-8077-M002
Marker: IgG (H&L)
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Rabbit
Conjugation: Biotin
Size: 2 mg

Catalog No. 50-000-87336


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.