Learn More
Immunology Consultants Affinity Purified Rabbit anti-C-Myc Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants RMYC45A
Encompass_Preferred
Rabbits were immunized with highly purified EQKLISEEDL conjugated with KLH (C-MYC) and the resulting antiserum was collected. Antibody was solid phase adsorbed then immunoaffinity purified off an antigen containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with EQKLISEEDL as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2062