Learn More
Immunology Consultants HRP Conjugated Rabbit anti-DYKDDDDK (FLAG) Antibody, 0.1 mg
SDP
Supplier:  Immunology Consultants RFLG45PZ
Encompass_Preferred
Rabbits were immunized with highly purified DYKDDDDK epitope conjugated to KLH and the resulting antiserum was collected. Antibody was immunoaffinity purified off a peptide containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with DYKDDDDK as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-196-2083