Learn More
Enzo Life Sciences Rabbit anti-rat IgG, polyclonal antibody (HRP conjugate) (1ml)
SDP
Catalog No. 502007394
Encompass_Preferred
Host: Rabbit. Immunogen: Purified rat IgG. Species reactivity: Rat. Specificity: Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule rat IgG. It also reacts with the light chains of other rat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Applications: ELISA, IHC, WB. Recommended Dilutions: ELISA (1:2,500-50,000) Immunohistochemistry (1:250-2500) Western Blot (1:5,000-100,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In 0.005M sodium phosphate, pH 7.6, containing 0.125M sodium chloride, 7.5mg/mL BSA (IgG-free, protease-free), and 50% glycerol.
Catalog No. 50-200-7394 Supplier Enzo Life Sciences Supplier No. ADISAB200J
Host: Rabbit. Immunogen: Purified rat IgG. Species reactivity: Rat. Specificity: Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule rat IgG. It also reacts with the light chains of other rat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Applications: ELISA, IHC, WB. Recommended Dilutions: ELISA (1:2,500-50,000) Immunohistochemistry (1:250-2500) Western Blot (1:5,000-100,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In 0.005M sodium phosphate, pH 7.6, containing 0.125M sodium chloride, 7.5mg/mL BSA (IgG-free, protease-free), and 50% glycerol.