Learn More
Bellco Glass, Inc. Round Bottom Canister Only,38x135mm
SDP
Catalog No. 501944657
Encompass_Preferred
ROUND BOTTOM GLASS PIPETTE CANISTER. MANUFACTURED FROM TYPE 1, CLASS A BOROSILICATE GLASS COMPONENTS WILL WITHSTAND REPEATED DRY OR STEAM HEAT STERILIZATION CASE Qty: 6
Catalog No. 50-194-4657 Supplier Bellco Glass, Inc. Supplier No. 128700135
ROUND BOTTOM GLASS PIPETTE CANISTER. MANUFACTURED FROM TYPE 1, CLASS A BOROSILICATE GLASS COMPONENTS WILL WITHSTAND REPEATED DRY OR STEAM HEAT STERILIZATION CASE Qty: 6