Learn More
Genscript Corporation MonoRab™ Rabbit IgG Control [PE] (Whole Molecule), mAb
Supplier:  Genscript Corporation A02025200
Encompass
Rabbit IgG Control is suitable for use as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, western blot, immunoprecipitation, immunodiffusion, and immunoelectrophoresis, flow cytometry and immunofluorescence. GenScript MonoRab™ Rabbit IgG Control [PE] (Whole Molecule), mAb is PE conjugated MonoRab™ Rabbit IgG Control (Whole Molecule), mAb.
Catalog No. NC1979782