Learn More
Biocare Medical, LLC RB-ON-CANINE HRP-POLYMER 100ML
SDP
Supplier:  Biocare Medical, LLC RC542L
Encompass_Preferred
Rabbit-on-Canine HRP-Polymer is designed as a one-step detection for rabbit antibodies on canine and feline tissues. Combine together for universal or simultaneous double stains. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-832-70