Learn More
Biocare Medical, LLC RB-ON-CANINE HRP-POLYMER 25ML
SDP
Catalog No. 5083269
Encompass_Preferred
Rabbit-on-Canine HRP-Polymer is designed as a one-step detection for rabbit antibodies on canine and feline tissues. Combine together for universal or simultaneous double stains. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-832-69 Supplier Biocare Medical, LLC Supplier No. RC542H
Rabbit-on-Canine HRP-Polymer is designed as a one-step detection for rabbit antibodies on canine and feline tissues. Combine together for universal or simultaneous double stains. It is labeled with horseradish peroxidase (HRP).