Learn More
Biocare Medical, LLC RB-ON-RODENT AP-POLYMER 25ML
SDP
Catalog No. 5083282
Encompass_Preferred
The Rabbit-on-Rodent AP-Polymer is for use of rabbit primary antibodies on mouse or rat tissues. Rabbit antibodies display minimum cross-reactivity to endogenous rodent IgG. It is labeled with alkaline phosphatase (AP).
Catalog No. 50-832-82 Supplier Biocare Medical, LLC Supplier No. RMR625H
The Rabbit-on-Rodent AP-Polymer is for use of rabbit primary antibodies on mouse or rat tissues. Rabbit antibodies display minimum cross-reactivity to endogenous rodent IgG. It is labeled with alkaline phosphatase (AP).