Learn More
Biocare Medical, LLC Rabbit-on-Rodent HRP-Polymer, 6ml
SDP
Catalog No. 5083278
Encompass_Preferred
The Rabbit-on-Rodent HRP-Polymer is for use of rabbit primary antibodies on mouse or rat tissues. Rabbit antibodies display minimum cross-reactivity to endogenous rodent IgG. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-832-78 Supplier Biocare Medical, LLC Supplier No. RMR622G
The Rabbit-on-Rodent HRP-Polymer is for use of rabbit primary antibodies on mouse or rat tissues. Rabbit antibodies display minimum cross-reactivity to endogenous rodent IgG. It is labeled with horseradish peroxidase (HRP).