Learn More
Medchemexpress LLC Rb-opd (NO-red) | 1034863-51-8 | 95.0% | 532.68 | C34H36N4O2 | 10 MG
SDP
Catalog No. 5000297793
Encompass_Preferred
RB-OPD (NO-red) is an o-phenylenediamine (OPD)-locked rhodamine-based fluorescent probe for selective and sensitive detection of nitric oxide in biochemical assays and imaging. It has excitation/emission maxima at 550 nm and 590 nm and is supplied as a solid or as a DMSO solution for research use only.
  • Selective detection of nitric oxide via spiro ring opening mechanism.
  • Excitation/emission: 550 nm/590 nm, compatible with common fluorescence setups.
  • High reported purity (95.0%), suitable for research assays.
  • Available as solid (small mg quantities) or as a 10 mM solution in dimethyl sulfoxide (DMSO).
  • Compact quantities support small-scale imaging and assay workflows.
Catalog No. 50-002-97793 Supplier Medchemexpress LLC Supplier No. HYD126610MG
RB-OPD (NO-red) is an o-phenylenediamine (OPD)-locked rhodamine-based fluorescent probe for selective and sensitive detection of nitric oxide in biochemical assays and imaging. It has excitation/emission maxima at 550 nm and 590 nm and is supplied as a solid or as a DMSO solution for research use only.
  • Selective detection of nitric oxide via spiro ring opening mechanism.
  • Excitation/emission: 550 nm/590 nm, compatible with common fluorescence setups.
  • High reported purity (95.0%), suitable for research assays.
  • Available as solid (small mg quantities) or as a 10 mM solution in dimethyl sulfoxide (DMSO).
  • Compact quantities support small-scale imaging and assay workflows.