Learn More
Enzo Life Sciences Retinoblastoma protein monoclonal antibody (MAb245) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA1880100
Encompass_Preferred
Clone: mAb245. Isotype: IgG1. Species reactivity: Human Primate. Specificity: Recognizes underphosphorylated and highly phosphorylated Rb. Detects an epitope between aa 332-344 of human Rb. Applications: WB. Formulation: Liquid. In 10 mM PBS, pH 7.2, 150 mM NaCl, 0.09% sodium azide.
Catalog No. 50-201-1765