Learn More
Innovative Research Inc Rb Vtrnctn Ply Frctn Btn lbl
Catalog No. 502744073
Encompass_Preferred
Rabbit Anti Human Vitronectin Polyclonal Fractionated Biotin Labeled 1mg - Rabbit Anti Human Vitronectin Polyclonal Antiserum from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antiserum is whole serum containing polyclonal antibodies (predominantly IgG & IgM) directed against the specified antigen and includes host antibodies and immunoglobulins of all classes along with other serum proteins. This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of paraffin sections and Immunohistochemistry of PFA fixed sections. Please note this product cross reacts with bovine vitronectin.
Catalog No. 50-274-4073 Supplier Innovative Research Inc Supplier No. IRBAHUVTNGFBC1MG
Rabbit Anti Human Vitronectin Polyclonal Fractionated Biotin Labeled 1mg - Rabbit Anti Human Vitronectin Polyclonal Antiserum from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antiserum is whole serum containing polyclonal antibodies (predominantly IgG & IgM) directed against the specified antigen and includes host antibodies and immunoglobulins of all classes along with other serum proteins. This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of paraffin sections and Immunohistochemistry of PFA fixed sections. Please note this product cross reacts with bovine vitronectin.