Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5925MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determined via Mass Spec. Conjugate/Tag: Unconjugated. Clone: 1F8. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa, A431 or BT-20 cells. Breast or Colon carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Rb1 protein fragment of 283 amino acid residues. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 105kDa.Chromosome: 13q14.2. Gene Symbol: RB1. Gene ID: 5925. Ensemble: ENSG00000139687. Uniprot: NP_000312.
Catalog No. 50-188-5124