Learn More
Aviva Systems Biology Corp RB1 Antibody - N-terminal region (ARP58065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RB1. It was validated on Western Blot. RB1,retinoblastoma 1,RB; pRb; OSRC; pp110; p105-Rb; RB1,retinoblastoma-associated protein,retinoblastoma-associated protein,retinoblastoma suspectibility proteinThe immunogen is a synthetic peptide directed towards the N terminal region of human RB1. Alternate names: OSRC, RB, pRb, pp110, p105-RbApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-433