Learn More
MyBioSource RB1 Recombinant Protein, Retinoblastoma, 1mg
SDP
Supplier:  MyBioSource MBS1432541MG
Encompass_Preferred
The product is a recombinant human retinoblastoma protein (RB1) expressed in E. coli with a molecular weight of 110 kDa. It is supplied as a lyophilized powder containing 1 mg of protein that can be reconstituted in water. The RB1 protein plays a key role as a tumor suppressor by regulating cell proliferation. This recombinant protein can be used for studying RB1 interactions and developing assays. It is suitable for in vitro experiments in biomedical research.
Catalog No. NC2819379