Learn More
Aviva Systems Biology Corp RB1CC1 Antibody - C-terminal region (ARP50467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RB1CC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RB1CC1. Alternate names: CC1, DRAGOU14, FIP200ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60351