Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB670 Mouse Anti-Rat IgG1, IgG2a, BD OptiBuild™

Catalog No. BDB772252 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772252 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772252 Supplier BD Biosciences Supplier No. 772252
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Rat Samples. It is validated for Flow Cytometry.

The G28-5 monoclonal antibody reacts specifically with rat IgG1 and IgG2a. It does not react with other Ig isotypes. The BD Horizon RealBlueTM 670 (RB670) Dye is a tandem fluorochrome with an excitation maximum at 492 nm and an emission maximum at 670 nm. It can be used on both spectral and conventional cytometers and is designed to be primarily excited by the Blue laser (488-nm).
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G28-5
Concentration 0.2 mg/mL
Conjugate RB670
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IgG1/IgG2a; Ig gamma-1/2a; Immunoglobulin gamma 1/2a
Host Species Mouse
Immunogen Pooled Rat IgG
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 299354
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.