Learn More
Description
Specifications
Specifications
| Concentration | 0.1 mg/mL |
| Conjugate | RB670 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Content And Storage | Streptavidin was conjugated with dye under optimum conditions, and unconjugated Streptavidin and free dye were removed Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.