Learn More
Aviva Systems Biology Corp RBAK Antibody - middle region (ARP32972_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32972_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBAK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBAK. Alternate names: ZNF769ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54837