Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology RBAP46/RBAP48 Monoclonal Antibody, Rabbit IgG, Western Blotting, 100 μl
SDP

Catalog No. 502730200
Encompass

This rabbit monoclonal antibody recognizes endogenous levels of total RBAP46 and RBAP48 proteins. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of human RBAP48 protein. RBAP46 and RBAP48 are components of several protein complexes involved in chromatin regulation, including CAF-1, HAT1, NURF, NuRD, Sin3/HDAC, and PRC2.

  • Validated for Western blotting
  • Reactivity: human, mouse, rat, and monkey
  • Predicted reactivity: hamster, chicken, guinea pig, and rabbit
  • Target molecular weight: 48 kDa
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide

Catalog No. 50-273-0200 Supplier Cell Signaling Technology Supplier No. 9067S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit monoclonal antibody recognizes endogenous levels of total RBAP46 and RBAP48 proteins. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of human RBAP48 protein. RBAP46 and RBAP48 are components of several protein complexes involved in chromatin regulation, including CAF-1, HAT1, NURF, NuRD, Sin3/HDAC, and PRC2.

  • Validated for Western blotting
  • Reactivity: human, mouse, rat, and monkey
  • Predicted reactivity: hamster, chicken, guinea pig, and rabbit
  • Target molecular weight: 48 kDa
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.