Learn More
Biomatik Corporation Rbbp4 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07067
Encompass_Preferred
Rbbp4 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to RBBP4. The Rbbp4 Recombinant Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Rbap48. Applications: ELISA, WB, IHC, IF. Target Synonyms: RBBP4. Accession Number: Q09028. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7104