Learn More
Sino Biological Recombinant Human PRMT3 Protein (Full Length, GST Tag), Active
Catalog No. 502573705
Encompass_Preferred
Recombinant full length human PRMT3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3705 Supplier Sino Biological Supplier No. P365380CG50
Recombinant full length human PRMT3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.