Learn More
ABclonal Technology RBBP5 Rabbit mAb
SDP
Catalog No. 5000005067
Encompass_Preferred
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins The encoded protein binds directly to retinoblastoma protein which regulates cell proliferation It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene
Catalog No. 50-000-05067 Supplier ABclonal Technology Supplier No. A2312420UL
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins The encoded protein binds directly to retinoblastoma protein which regulates cell proliferation It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene