Learn More
Aviva Systems Biology Corp RBBP8 Antibody - C-terminal region (ARP50887_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50887_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBBP8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RBBP8. Alternate names: CTIP, RIM, JWDS, SAE2, SCKL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-54070