Learn More
Aviva Systems Biology Corp RBBP8 Antibody - middle region (ARP50888_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50888_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBBP8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RBBP8. Alternate names: CTIP, RIM, JWDS, SAE2, SCKL2ApplicationWB. Species reactivity: Human
Catalog No. 50-285-548