Learn More
ENG Scientific RBC Diluting Fluid (Hayem™'s Solution)
Catalog No. ES5601
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES5601 1 x 100 mL
ES5600 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Do not put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. ES5601 Supplier ENG Scientific Supplier No. 5601
Add to Cart
Add to Cart
The products indicated contain mercury. Do not put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
RBC Diluting Fluid (Hayem™'s Solution) is used for diluting RBC's. Contains Sodium Sulfate, Sodium Chloride and Mercuric Chloride. Settling time is between 1 to 3 minutes.
  • RBC Diluting Fluid (Hayem™'s Solution)
  • Contents: Sodium Sulfate, Sodium Chloride, Mercuric Chloride
  • For diluting Blood prior to counting red Blood cells with a hemocytometer
  • Staining time 1 - 3 minutes
  • For in-vitro diagnostic use only
Specifications
Quantity 1 x 100 mL
For Use With (Application) For diluting Blood prior to counting red Blood cells with a hemocytometer
Staining Time 1 to 3 min.
Packaging Bottle
Product Type Diluting Fluid
For in vitro diagnostic use only.