Learn More
ENG Scientific RBC Diluting Fluid (Hayem™'s Solution)
Description
- RBC Diluting Fluid (Hayem™'s Solution)
- Contents: Sodium Sulfate, Sodium Chloride, Mercuric Chloride
- For diluting Blood prior to counting red Blood cells with a hemocytometer
- Staining time 1 - 3 minutes
- For in-vitro diagnostic use only
Specifications
Specifications
| Quantity | 1 x 100 mL |
| For Use With (Application) | For diluting Blood prior to counting red Blood cells with a hemocytometer |
| Staining Time | 1 to 3 min. |
| Packaging | Bottle |
For in vitro diagnostic use only.