Learn More
Zymo Research Corporation RBC Lysis Buffer, 100 ml
SDP
Catalog No. 502037234
Encompass_Preferred
RBC Lysis Buffer used for red blood cell lysis for RNA extraction from pelleted blood cells in the Quick-RNA Whole Blood kit.
Catalog No. 50-203-7234 Supplier Zymo Research Corporation Supplier No. R10222100
RBC Lysis Buffer used for red blood cell lysis for RNA extraction from pelleted blood cells in the Quick-RNA Whole Blood kit.